siRNA Details
Gene Symbol: Phrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139093.1 | NP_620793.1 | 8 | 949 | ||
XM_006230510.2 | XP_006230572.1 | 9 | 1117 | |||
XM_006230511.2 | XP_006230573.1 | 9 | 1075 | |||
XM_006230513.2 | XP_006230575.1 | 9 | 1114 | |||
XM_006230514.2 | XP_006230576.1 | 9 | 1117 | |||
XM_006230515.2 | XP_006230577.1 | 9 | 1117 | |||
XM_006230516.2 | XP_006230578.1 | 9 | 1117 | |||
XM_006230517.2 | XP_006230579.1 | 9 | 1114 | |||
GenBank mRNA | U49057.1 | - | 8 | 949 |
