siRNA Details
Gene Symbol: Nos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052799.1 | NP_434686.1 | 21 | 3333 | ||
XM_008769210.1 | XP_008767432.1 | 22 | 4144 | |||
GenBank mRNA | U67309.1 | - | 21 | 3333 | ||
X59949.1 | - | 20 | 3478 |
