siRNA Details
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012613.1 | NP_036745.1 | 5, 6 | 1623 | ||
XM_006232591.2 | XP_006232653.1 | 6, 7 | 2124 | |||
GenBank mRNA | BC128742.1 | - | 5, 6 | 1609 | 7458324 | |
M74535.1 | - | 6, 7 | 1572 | |||
X14773.1 | - | 5, 6 | 1480 |
