siRNA Details
Gene Symbol: Atrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773366.1 | XP_008771588.1 | 10 | 3349 | ||
GenBank mRNA | D64059.1 | - | 1 | 481 |
Gene Symbol: LOC103690008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257157.2 | XP_006257219.2 | 3689 | |||
XM_008773351.1 | XP_008771573.1 | 3686 | ||||
XM_008773352.1 | XP_008771574.1 | 7 | 3575 | |||
XM_008773353.1 | XP_008771575.1 | 7 | 3572 |
