siRNA Details
Gene Symbol: Prkca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105713.1 | NP_001099183.1 | 4, 5 | 673 | ||
XM_008768310.1 | XP_008766532.1 | 4, 5 | 910 | |||
GenBank mRNA | BC169007.1 | - | 4, 5 | 622 | 7387040 | |
X07286.1 | - | 4, 5 | 624 |
