siRNA Details
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012637.2 | NP_036769.1 | 2 | 178 | ||
XM_006235638.2 | XP_006235700.1 | 2 | 247 | |||
XM_006235639.2 | XP_006235701.1 | 2 | 248 | |||
GenBank mRNA | BC081829.1 | - | 2 | 178 | 7135855 | |
M33962.1 | - | 2 | 220 |
