siRNA Details
Gene Symbol: Sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012655.2 | NP_036787.2 | 6 | 2381 | ||
GenBank mRNA | AY305388.1 | - | 1 | 77 | ||
D12768.1 | - | 6 | 2397 |

More Information
Other Species Targeted

Gene Ontology Categories:
