siRNA Details
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012687.1 | NP_036819.1 | 11 | 1417 | ||
XM_008762759.1 | XP_008760981.1 | 11 | 1478 | |||
XM_008762760.1 | XP_008760982.1 | 9 | 2875 | |||
GenBank mRNA | D28773.1 | - | 11 | 1417 | ||
D31798.1 | - | 11 | 1339 |
