siRNA Details
Gene Symbol: Slc22a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012697.1 | NP_036829.1 | 5 | 930 | ||
GenBank mRNA | BC078883.1 | - | 4 | 804 | 7130305 | |
U76379.1 | - | 4 | 826 | |||
X78855.1 | - | 5 | 930 |
