siRNA Details
Gene Symbol: Klrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133512.2 | NP_598196.1 | 6 | 495 | ||
XM_006237073.2 | XP_006237135.1 | 6 | 1677 | |||
XM_006237074.2 | XP_006237136.1 | 6 | 576 | |||
XM_006237075.2 | XP_006237137.1 | 6 | 645 | |||
XM_006237076.2 | XP_006237138.1 | 6 | 483 | |||
XM_006237077.2 | XP_006237139.1 | 6 | 533 | |||
GenBank mRNA | AF009511.1 | - | 5 | 449 | ||
FQ241294.1 | - | 6 | 495 |
