siRNA Details
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030991.3 | NP_112253.1 | 375 | |||
GenBank mRNA | AB003992.1 | - | 166 | |||
AF245227.1 | - | 374 | ||||
BC087699.1 | - | 330 | 7314318 | |||
FQ212040.1 | - | 346 | ||||
FQ213289.1 | - | 4 | 343 | |||
FQ213923.1 | - | 346 | ||||
FQ214162.1 | - | 375 | ||||
U56262.1 | - | 139 |
