siRNA Details
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017075.2 | NP_058771.2 | 3 | 280 | ||
GenBank mRNA | D00512.1 | - | 3 | 212 | ||
D13921.1 | - | 3 | 212 | |||
FM065829.1 | - | 3 | 280 | |||
FM071064.1 | - | 1 | 68 | |||
FQ217320.1 | - | 3 | 218 |
