siRNA Details
Gene Symbol: Adm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012715.1 | NP_036847.1 | 4 | 433 | ||
XM_008759687.1 | XP_008757909.1 | 5 | 1940 | |||
GenBank mRNA | BC061775.1 | - | 4 | 429 | 5621783 | |
D15069.1 | - | 4 | 415 | |||
U15419.1 | - | 4 | 433 |
