siRNA Details
Gene Symbol: Sds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053962.3 | NP_446414.3 | 9 | 1097 | ||
XM_008769213.1 | XP_008767435.1 | 8 | 1595 | |||
GenBank mRNA | BC088110.1 | - | 9 | 1097 | 7323391 | |
J03863.1 | - | 9 | 1295 | |||
Y00752.1 | - | 9 | 1114 |
