siRNA Details
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163168.2 | NP_001156640.1 | 12 | 2008 | ||
NM_012731.2 | NP_036863.1 | 12 | 1993 | |||
XM_006253507.2 | XP_006253569.1 | 12 | 1944 | |||
XM_008771425.1 | XP_008769647.1 | 12 | 1949 | |||
XM_008771426.1 | XP_008769648.1 | 12 | 1835 | |||
XM_008771427.1 | XP_008769649.1 | 12 | 1943 | |||
GenBank mRNA | AY265419.1 | - | 12 | 1773 | ||
M55291.1 | - | 12 | 1996 | |||
M55292.1 | - | 12 | 1773 | |||
M55293.1 | - | 11 | 1440 |
