siRNA Details
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163168.2 | NP_001156640.1 | 3, 4 | 971 | ||
NM_012731.2 | NP_036863.1 | 3, 4 | 956 | |||
XM_006253507.2 | XP_006253569.1 | 3, 4 | 907 | |||
XM_008771425.1 | XP_008769647.1 | 3, 4 | 912 | |||
XM_008771426.1 | XP_008769648.1 | 3, 4 | 798 | |||
XM_008771427.1 | XP_008769649.1 | 3, 4 | 906 | |||
GenBank mRNA | AY265419.1 | - | 3, 4 | 736 | ||
M55291.1 | - | 3, 4 | 959 | |||
M55292.1 | - | 3, 4 | 736 | |||
M55293.1 | - | 2, 3 | 403 |
