siRNA Details
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163168.2 | NP_001156640.1 | 8 | 1413 | ||
NM_012731.2 | NP_036863.1 | 8 | 1398 | |||
XM_006253507.2 | XP_006253569.1 | 8 | 1349 | |||
XM_008771425.1 | XP_008769647.1 | 8 | 1354 | |||
XM_008771426.1 | XP_008769648.1 | 8 | 1240 | |||
XM_008771427.1 | XP_008769649.1 | 8 | 1348 | |||
GenBank mRNA | AY265419.1 | - | 8 | 1178 | ||
KC855568.1 | - | 2 | 102 | |||
M55291.1 | - | 8 | 1401 | |||
M55292.1 | - | 8 | 1178 | |||
M55293.1 | - | 7 | 845 |
