siRNA Details
Gene Symbol: Hk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022179.2 | NP_071515.1 | 16 | 2156 | ||
XM_006253602.2 | XP_006253664.1 | 17 | 2371 | |||
XM_006253603.2 | XP_006253665.1 | 16 | 2351 | |||
XM_008771484.1 | XP_008769706.1 | 16 | 2420 | |||
GenBank mRNA | BC100648.1 | - | 16 | 2133 | 7932574 |
