siRNA Details
Gene Symbol: Scarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031541.1 | NP_113729.1 | 1 | 279 | ||
GenBank mRNA | AB002151.1 | - | 1 | 279 | ||
AY451993.1 | - | 1 | 184 | |||
AY682847.1 | - | 1 | 184 | |||
BC076504.1 | - | 1 | 188 | 7099863 | ||
D89655.1 | - | 1 | 197 | |||
U76205.1 | - | 1 | 125 |
