siRNA Details
Gene Symbol: Fgfr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109904.1 | NP_001103374.1 | 15 | 2150 | ||
XM_006253606.2 | XP_006253668.1 | 15 | 2167 | |||
XM_008771485.1 | XP_008769707.1 | 15 | 2161 | |||
GenBank mRNA | BC100260.1 | - | 14 | 2242 | 7100540 | |
M91599.1 | - | 12 | 1616 |
