siRNA Details
Gene Symbol: Mr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100635.1 | NP_001094105.1 | 3 | 572 | ||
XM_006250030.2 | XP_006250092.1 | 3 | 686 | |||
XM_008769606.1 | XP_008767828.1 | 3 | 686 | |||
GenBank mRNA | Y13972.1 | - | 2 | 497 |
