siRNA Details
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012751.1 | NP_036883.1 | 4 | 544 | ||
XM_006246596.2 | XP_006246658.1 | 5 | 517 | |||
GenBank mRNA | BC085757.1 | - | 4 | 589 | 7192278 | |
D28561.1 | - | 4 | 544 | |||
FQ216450.1 | - | 5 | 483 | |||
FQ232657.1 | - | 4 | 603 | |||
FQ233419.1 | - | 4 | 579 | |||
J04524.1 | - | 4 | 526 | |||
KJ696745.1 | - | 4 | 429 | |||
M25482.1 | - | 4 | 509 | |||
X14771.1 | - | 4 | 557 |
