siRNA Details
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012762.2 | NP_036894.2 | 6 | 949 | ||
GenBank mRNA | D85899.1 | - | 6 | 949 | ||
FQ228818.1 | - | 4 | 582 | |||
S79676.1 | - | 2 | 292 | |||
U14647.1 | - | 6 | 949 | |||
U34621.1 | - | 2 | 268 |
