siRNA Details
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012762.2 | NP_036894.2 | 4 | 548 | ||
GenBank mRNA | D85899.1 | - | 4 | 548 | ||
FQ228818.1 | - | 2 | 180 | |||
U14647.1 | - | 4 | 548 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012762.2 | NP_036894.2 | 4 | 548 | ||
GenBank mRNA | D85899.1 | - | 4 | 548 | ||
FQ228818.1 | - | 2 | 180 | |||
U14647.1 | - | 4 | 548 |