siRNA Details
Gene Symbol: Gdi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017088.2 | NP_058784.2 | 2, 3 | 278 | ||
GenBank mRNA | BC091156.1 | - | 2, 3 | 282 | ||
BC099763.1 | - | 2, 3 | 278 | 7930350 | ||
U07952.1 | - | 2, 3 | 195 | |||
X74402.1 | - | 2, 3 | 184 |
