siRNA Details
Gene Symbol: Arsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033443.1 | NP_254278.1 | 7 | 1344 | ||
GenBank mRNA | BC127450.1 | - | 7 | 1235 | ||
BC168241.1 | - | 1315 | 7934677 | |||
CF113828.1 | - | 4 | 512 | |||
D49434.1 | - | 7 | 1086 |
