siRNA Details
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012789.1 | NP_036921.1 | 9, 10 | 866 | ||
XM_008761879.1 | XP_008760101.1 | 10, 11 | 1375 | |||
GenBank mRNA | J02997.1 | - | 9, 10 | 806 | ||
J04591.1 | - | 9, 10 | 866 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010074, NM_001159543, AK164775, AK085370, BC022183, and others.
Gene Ontology Categories:
