siRNA Details
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012789.1 | NP_036921.1 | 14 | 1306 | ||
XM_008761879.1 | XP_008760101.1 | 15 | 1815 | |||
GenBank mRNA | J02997.1 | - | 14 | 1246 | ||
J04591.1 | - | 14 | 1306 |

More Information
Other Species Targeted

Gene Ontology Categories:
