siRNA Details
Gene Symbol: Itpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007235.2 | NP_001007236.2 | 32 | 4150 | ||
NM_001270596.1 | NP_001257525.1 | 31 | 4105 | |||
NM_001270597.1 | NP_001257526.1 | 32 | 4150 | |||
XM_006236987.2 | XP_006237049.1 | 32 | 4215 | |||
XM_008763167.1 | XP_008761389.1 | 32 | 4613 | |||
XM_008763168.1 | XP_008761390.1 | 32 | 4613 | |||
XM_008763169.1 | XP_008761391.1 | 32 | 4586 | |||
XM_008763170.1 | XP_008761392.1 | 32 | 4586 | |||
XM_008763171.1 | XP_008761393.1 | 32 | 4612 | |||
XM_008763172.1 | XP_008761394.1 | 32 | 4612 | |||
XM_008763173.1 | XP_008761395.1 | 31 | 4567 | |||
XM_008763174.1 | XP_008761396.1 | 32 | 4612 | |||
XM_008763175.1 | XP_008761397.1 | 31 | 4540 | |||
XM_008763176.1 | XP_008761398.1 | 32 | 4611 | |||
XM_008763177.1 | XP_008761399.1 | 31 | 4567 | |||
XM_008763178.1 | XP_008761400.1 | 31 | 4539 | |||
GenBank mRNA | GQ233032.1 | - | 30 | 3912 | ||
J05510.1 | - | 32 | 4241 |
