siRNA Details
Gene Symbol: Mapk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270556.1 | NP_001257485.1 | 8 | 1182 | ||
NM_012806.2 | NP_036938.2 | 8 | 1209 | |||
XM_008769992.1 | XP_008768214.1 | 7 | 4001 | |||
GenBank mRNA | DQ377224.1 | - | 5 | 556 | ||
FQ211777.1 | - | 8 | 1181 | |||
L27128.1 | - | 8 | 919 |
