siRNA Details
Gene Symbol: Nup153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100470.1 | NP_001093940.1 | 22 | 4901 | ||
XM_008771532.1 | XP_008769754.1 | 21 | 4684 | |||
GenBank mRNA | BC084700.1 | - | 5 | 1690 | ||
CO574678.1 | - | 2 | 116 | |||
L06821.1 | - | 22 | 4901 |

More Information
Other Species Targeted

Gene Ontology Categories:
