siRNA Details
Gene Symbol: Nup153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100470.1 | NP_001093940.1 | 16 | 2865 | ||
XM_008771532.1 | XP_008769754.1 | 15 | 2648 | |||
GenBank mRNA | L06821.1 | - | 16 | 2865 |
