siRNA Details
Gene Symbol: Amacr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012816.2 | NP_036948.2 | 5 | 783 | ||
GenBank mRNA | AY325250.1 | - | 1032 | |||
BC078885.1 | - | 5 | 751 | 7110324 | ||
BC091163.1 | - | 5 | 752 | 7130642 | ||
BC127505.1 | - | 5 | 772 | 7325347 | ||
U89905.1 | - | 5 | 771 | |||
Y08172.1 | - | 5 | 761 |
