siRNA Details
Gene Symbol: Acsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012820.1 | NP_036952.1 | 6 | 641 | ||
XM_006253122.1 | XP_006253184.1 | 7 | 1301 | |||
XM_006253123.1 | XP_006253185.1 | 7 | 842 | |||
XM_006253124.2 | XP_006253186.1 | 7 | 761 | |||
XM_006253125.2 | XP_006253187.1 | 7 | 921 | |||
XM_006253126.1 | XP_006253188.1 | 7 | 767 | |||
XM_006253127.1 | XP_006253189.1 | 7 | 732 | |||
XM_008771254.1 | XP_008769476.1 | 6 | 667 | |||
GenBank mRNA | D90109.1 | - | 6 | 641 | ||
FQ217275.1 | - | 6 | 611 |
