siRNA Details
Gene Symbol: Anxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012823.2 | NP_036955.2 | 5 | 376 | ||
XM_006250694.2 | XP_006250756.1 | 5 | 740 | |||
XM_006250696.2 | XP_006250758.1 | 6 | 496 | |||
GenBank mRNA | BC081856.1 | - | 4 | 288 | 7191216 | |
FQ232970.1 | - | 5 | 367 | |||
M20559.1 | - | 5 | 324 |
