siRNA Details
Gene Symbol: Phtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191102.1 | NP_001178031.1 | 1 | 36 | ||
XM_006233061.2 | XP_006233123.1 | 2 | 2208 | |||
XM_006233063.2 | XP_006233125.1 | 2 | 2208 | |||
XM_006233064.2 | XP_006233126.1 | 2 | 2207 | |||
XM_006233065.2 | XP_006233127.1 | 2 | 2208 | |||
GenBank mRNA | AJ437403.1 | - | 1 | 42 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001163469, NM_001163468, NM_001163467, NM_013629, AK088714, and others.
Gene Ontology Categories:
