siRNA Details
Gene Symbol: Bmp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012827.2 | NP_036959.2 | 150 | |||
XM_006251746.2 | XP_006251808.1 | 912 | ||||
XM_006251747.2 | XP_006251809.1 | 462 | ||||
XM_006251748.2 | XP_006251810.1 | 3 | 444 | |||
XM_006251749.2 | XP_006251811.1 | 186 | ||||
XM_006251751.2 | XP_006251813.1 | 163 | ||||
XM_008770501.1 | XP_008768723.1 | 276 | ||||
GenBank mRNA | AY184241.1 | - | 1 | 12 | ||
BC078901.1 | - | 150 | 7121295 | |||
Z22607.1 | - | 3 | 412 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007554, AK145359, BC034053, BC052846, AK136527, and others.
Gene Ontology Categories:
