siRNA Details
Gene Symbol: Comp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012834.1 | NP_036966.1 | 10, 11 | 1131 | ||
XM_006252822.2 | XP_006252884.1 | 7, 8 | 1335 | |||
GenBank mRNA | X72914.1 | - | 10, 11 | 1131 |
