siRNA Details
Gene Symbol: Il10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012854.2 | NP_036986.2 | 1 | 142 | ||
XM_006249712.2 | XP_006249774.1 | 1 | 481 | |||
XM_008769426.1 | XP_008767648.1 | 1 | 481 | |||
GenBank mRNA | L02926.1 | - | 1 | 142 | ||
X60675.1 | - | 185 |
