siRNA Details
Gene Symbol: Lamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012857.2 | NP_036989.1 | 8 | 1029 | ||
GenBank mRNA | FQ221686.1 | - | 3 | 159 | ||
M34959.1 | - | 8 | 1029 | |||
U75406.1 | - | 2 | 80 | |||
X14765.1 | - | 7 | 865 |
