siRNA Details
Gene Symbol: Lamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012857.2 | NP_036989.1 | 6, 7 | 938 | ||
GenBank mRNA | FQ221686.1 | - | 1, 2 | 68 | ||
M34959.1 | - | 6, 7 | 938 | |||
X14765.1 | - | 5, 6 | 774 |
