siRNA Details
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012859.1 | NP_036991.1 | 2 | 1086 | ||
XM_006228391.2 | XP_006228453.1 | 2 | 219 | |||
XM_006228393.2 | XP_006228455.1 | 2 | 470 | |||
XM_006228394.2 | XP_006228456.1 | 2 | 1797 | |||
XM_008758895.1 | XP_008757117.1 | 2 | 203 | |||
XM_008758896.1 | XP_008757118.1 | 2 | 363 | |||
GenBank mRNA | AY428844.1 | - | 2 | 169 | ||
BC078888.1 | - | 2 | 1069 | |||
U40001.1 | - | 2 | 1086 | |||
X51415.1 | - | 1 | 608 |
