siRNA Details
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012859.1 | NP_036991.1 | 5 | 1914 | ||
XM_006228391.2 | XP_006228453.1 | 5 | 1047 | |||
XM_006228393.2 | XP_006228455.1 | 5 | 1298 | |||
XM_006228394.2 | XP_006228456.1 | 5 | 2625 | |||
XM_008758895.1 | XP_008757117.1 | 5 | 1031 | |||
XM_008758896.1 | XP_008757118.1 | 5 | 1191 | |||
GenBank mRNA | U40001.1 | - | 5 | 1914 | ||
X51415.1 | - | 4 | 1436 |
