siRNA Details
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012891.2 | NP_037023.1 | 16, 17 | 1633 | ||
GenBank mRNA | BC087036.1 | - | 16, 17 | 1654 | 7193535 | |
D30647.1 | - | 16, 17 | 1635 | |||
FQ219329.1 | - | 16, 17 | 1678 |
