siRNA Details
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012895.3 | NP_037027.2 | 3, 4 | 202 | ||
XM_006251631.1 | XP_006251693.1 | 3, 4 | 316 | |||
GenBank mRNA | BC081712.1 | - | 3, 4 | 202 | 7193044 | |
U57042.1 | - | 2, 3 | 142 | |||
U90340.1 | - | 3, 4 | 205 |
