siRNA Details
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012895.3 | NP_037027.2 | 6 | 488 | ||
XM_006251631.1 | XP_006251693.1 | 6 | 602 | |||
GenBank mRNA | BC081712.1 | - | 6 | 488 | 7193044 | |
FQ218369.1 | - | 2 | 151 | |||
FQ225093.1 | - | 2 | 159 | |||
U57042.1 | - | 5 | 428 | |||
U90340.1 | - | 6 | 491 |
