siRNA Details
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031555.1 | NP_113743.1 | 2 | 389 | ||
XM_006235117.2 | XP_006235179.1 | 2 | 50 | |||
GenBank mRNA | AB003104.1 | - | 2 | 390 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031555.1 | NP_113743.1 | 2 | 389 | ||
XM_006235117.2 | XP_006235179.1 | 2 | 50 | |||
GenBank mRNA | AB003104.1 | - | 2 | 390 |