siRNA Details
Gene Symbol: Cacna1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012918.3 | NP_037050.2 | 44 | 6182 | ||
XM_008772347.1 | XP_008770569.1 | 44 | 6633 | |||
XM_008772348.1 | XP_008770570.1 | 44 | 6633 | |||
XM_008772349.1 | XP_008770571.1 | 44 | 6630 | |||
XM_008772350.1 | XP_008770572.1 | 43 | 6627 | |||
XM_008772351.1 | XP_008770573.1 | 43 | 6621 | |||
XM_008772353.1 | XP_008770575.1 | 45 | 6525 | |||
XM_008772354.1 | XP_008770576.1 | 45 | 6525 | |||
XM_008772355.1 | XP_008770577.1 | 44 | 6519 | |||
XM_008772356.1 | XP_008770578.1 | 44 | 6513 | |||
XM_008772357.1 | XP_008770579.1 | 43 | 6507 | |||
XM_008772358.1 | XP_008770580.1 | 43 | 6507 | |||
XM_008772360.1 | XP_008770582.1 | 44 | 6633 | |||
XM_008772361.1 | XP_008770583.1 | 47 | 6952 | |||
GenBank mRNA | AF051527.1 | - | 12 | 1196 | ||
M64373.1 | - | 44 | 6182 |
