siRNA Details
Gene Symbol: Cacna1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012918.3 | NP_037050.2 | 42, 43 | 6051 | ||
XM_008772347.1 | XP_008770569.1 | 42, 43 | 6502 | |||
XM_008772348.1 | XP_008770570.1 | 42, 43 | 6502 | |||
XM_008772349.1 | XP_008770571.1 | 42, 43 | 6499 | |||
XM_008772350.1 | XP_008770572.1 | 41, 42 | 6496 | |||
XM_008772351.1 | XP_008770573.1 | 41, 42 | 6490 | |||
XM_008772352.1 | XP_008770574.1 | 42, 43 | 6502 | |||
XM_008772353.1 | XP_008770575.1 | 43, 44 | 6394 | |||
XM_008772354.1 | XP_008770576.1 | 43, 44 | 6394 | |||
XM_008772355.1 | XP_008770577.1 | 42, 43 | 6388 | |||
XM_008772356.1 | XP_008770578.1 | 42, 43 | 6382 | |||
XM_008772357.1 | XP_008770579.1 | 41, 42 | 6376 | |||
XM_008772358.1 | XP_008770580.1 | 41, 42 | 6376 | |||
XM_008772359.1 | XP_008770581.1 | 43, 44 | 6394 | |||
XM_008772360.1 | XP_008770582.1 | 42, 43 | 6502 | |||
XM_008772361.1 | XP_008770583.1 | 45, 46 | 6821 | |||
GenBank mRNA | AF051526.1 | - | 13, 14 | 1356 | ||
AF051527.1 | - | 10, 11 | 1065 | |||
M64373.1 | - | 42, 43 | 6051 |
