siRNA Details
Gene Symbol: Alx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012921.1 | NP_037053.1 | 4 | 1412 | ||
XM_006241293.2 | XP_006241355.1 | 5 | 1224 | |||
GenBank mRNA | BC158591.1 | - | 4 | 1188 | 9023971 | |
L14018.1 | - | 4 | 1412 |
